Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603049 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TXNL1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
TXNL1 - thioredoxin like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004786.2 | 994 | Intron | NP_004777.1 | |||
XM_006722580.3 | 994 | Intron | XP_006722643.1 | |||
XM_011526263.2 | 994 | UTR 3 | XP_011524565.1 | |||
XM_011526264.2 | 994 | Intron | XP_011524566.1 | |||
XM_017026093.1 | 994 | Intron | XP_016881582.1 | |||
XM_017026094.1 | 994 | UTR 3 | XP_016881583.1 |
Set Membership: |
HapMap |