Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 169800 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CNDP2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CNDP2 - CNDP dipeptidase 2 (metallopeptidase M20 family) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001168499.1 | 84 | Intron | NP_001161971.1 | |||
NM_018235.2 | 84 | UTR 5 | NP_060705.2 | |||
XM_005266728.2 | 84 | UTR 5 | XP_005266785.1 | |||
XM_006722503.2 | 84 | Intron | XP_006722566.1 | |||
XM_011526071.2 | 84 | Intron | XP_011524373.1 | |||
XM_011526072.2 | 84 | Intron | XP_011524374.1 | |||
XM_011526073.2 | 84 | Intron | XP_011524375.1 |
Set Membership: |
HapMap |