Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
4 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606351 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ESPN PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ESPN - espin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_031475.2 | Intron | NP_113663.2 | ||||
XM_005263501.2 | Intron | XP_005263558.1 | ||||
XM_011542231.1 | Intron | XP_011540533.1 | ||||
XM_011542232.1 | Intron | XP_011540534.1 | ||||
XM_011542233.2 | Intron | XP_011540535.1 | ||||
XM_011542236.2 | Intron | XP_011540538.1 | ||||
XM_011542237.1 | Intron | XP_011540539.1 | ||||
XM_011542238.2 | Intron | XP_011540540.1 | ||||
XM_017002433.1 | Intron | XP_016857922.1 | ||||
XM_017002434.1 | Intron | XP_016857923.1 |
MIR4252 - microRNA 4252 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |