Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600385 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADCY7 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ADCY7 - adenylate cyclase 7 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
BRD7 - bromodomain containing 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001173984.2 | Intron | NP_001167455.1 | ||||
NM_013263.4 | Intron | NP_037395.2 | ||||
XM_011523046.2 | Intron | XP_011521348.1 | ||||
XM_011523047.2 | Intron | XP_011521349.1 | ||||
XM_011523048.2 | Intron | XP_011521350.1 | ||||
XM_011523049.2 | Intron | XP_011521351.1 | ||||
XM_011523050.2 | Intron | XP_011521352.1 | ||||
XM_017023179.1 | Intron | XP_016878668.1 | ||||
XM_017023180.1 | Intron | XP_016878669.1 | ||||
XM_017023181.1 | Intron | XP_016878670.1 |
Set Membership: |
HapMap |