Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
32 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614612 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KANK4 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
KANK4 - KN motif and ankyrin repeat domains 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320269.1 | Intron | NP_001307198.1 | ||||
NM_181712.4 | Intron | NP_859063.3 | ||||
XM_011540843.2 | Intron | XP_011539145.1 | ||||
XM_011540845.2 | Intron | XP_011539147.1 | ||||
XM_017000483.1 | Intron | XP_016855972.1 | ||||
XM_017000484.1 | Intron | XP_016855973.1 | ||||
XM_017000485.1 | Intron | XP_016855974.1 |
Set Membership: |
HapMap |