Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 164770 MIM: 173410 | ||||||||||||||||||||
Literature Links: |
CSF1R PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CSF1R - colony stimulating factor 1 receptor | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PDGFRB - platelet derived growth factor receptor beta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002609.3 | 5276 | UTR 3 | NP_002600.1 | |||
XM_005268464.2 | 5276 | UTR 3 | XP_005268521.1 | |||
XM_011537658.1 | 5276 | UTR 3 | XP_011535960.1 | |||
XM_011537659.1 | 5276 | UTR 3 | XP_011535961.1 |