Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300391 MIM: 300118 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
AMELX PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
AMELX - amelogenin, X-linked | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142.2 | 1897 | Intron | NP_001133.1 | |||
NM_182680.1 | 1897 | Intron | NP_872621.1 | |||
NM_182681.1 | 1897 | Intron | NP_872622.1 | |||
XM_017029404.1 | 1897 | UTR 3 | XP_016884893.1 |
ARHGAP6 - Rho GTPase activating protein 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001287242.1 | 1897 | Intron | NP_001274171.1 | |||
NM_006125.2 | 1897 | Intron | NP_006116.2 | |||
NM_013423.2 | 1897 | Intron | NP_038267.1 | |||
NM_013427.2 | 1897 | Intron | NP_038286.2 |
Set Membership: |
HapMap |