Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCDC83 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CCDC83 - coiled-coil domain containing 83 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286159.1 | Intron | NP_001273088.1 | ||||
NM_173556.4 | Intron | NP_775827.2 | ||||
XM_011544839.2 | Intron | XP_011543141.1 | ||||
XM_011544840.2 | Intron | XP_011543142.1 | ||||
XM_011544841.1 | Intron | XP_011543143.1 | ||||
XM_011544842.1 | Intron | XP_011543144.1 | ||||
XM_011544843.1 | Intron | XP_011543145.1 | ||||
XM_011544844.2 | Intron | XP_011543146.1 | ||||
XM_017017354.1 | Intron | XP_016872843.1 | ||||
XM_017017355.1 | Intron | XP_016872844.1 |