Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607243 MIM: 614766 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
AP4S1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
AP4S1 - adaptor related protein complex 4 sigma 1 subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001128126.2 | Intron | NP_001121598.1 | ||||
NM_001254726.1 | Intron | NP_001241655.1 | ||||
NM_001254727.1 | Intron | NP_001241656.1 | ||||
NM_001254728.1 | Intron | NP_001241657.1 | ||||
NM_001254729.1 | Intron | NP_001241658.1 | ||||
NM_007077.4 | Intron | NP_009008.2 | ||||
XM_005267293.4 | Intron | XP_005267350.1 | ||||
XM_011536371.2 | Intron | XP_011534673.1 | ||||
XM_011536372.2 | Intron | XP_011534674.1 |
STRN3 - striatin 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |