Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605855 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ATP10A PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ATP10A - ATPase phospholipid transporting 10A (putative) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024490.3 | 6059 | Intron | NP_077816.1 | |||
XM_005268261.4 | 6059 | Intron | XP_005268318.1 | |||
XM_011521826.2 | 6059 | Intron | XP_011520128.1 | |||
XM_011521828.2 | 6059 | UTR 3 | XP_011520130.1 | |||
XM_011521829.2 | 6059 | Intron | XP_011520131.1 | |||
XM_011521830.2 | 6059 | Intron | XP_011520132.2 | |||
XM_017022436.1 | 6059 | Intron | XP_016877925.1 | |||
XM_017022437.1 | 6059 | Intron | XP_016877926.1 |
Set Membership: |
HapMap |