Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CEP295 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CEP295 - centrosomal protein 295 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_033395.1 | Intron | NP_203753.1 | ||||
XM_005274366.1 | Intron | XP_005274423.1 | ||||
XM_005274367.2 | Intron | XP_005274424.1 | ||||
XM_005274368.2 | Intron | XP_005274425.1 | ||||
XM_011543047.1 | Intron | XP_011541349.1 | ||||
XM_011543048.1 | Intron | XP_011541350.1 | ||||
XM_011543049.1 | Intron | XP_011541351.1 | ||||
XM_011543050.2 | Intron | XP_011541352.1 | ||||
XM_011543053.2 | Intron | XP_011541355.1 | ||||
XM_011543054.2 | Intron | XP_011541356.1 | ||||
XM_017018470.1 | Intron | XP_016873959.1 | ||||
XM_017018471.1 | Intron | XP_016873960.1 | ||||
XM_017018472.1 | Intron | XP_016873961.1 | ||||
XM_017018473.1 | Intron | XP_016873962.1 | ||||
XM_017018474.1 | Intron | XP_016873963.1 | ||||
XM_017018475.1 | Intron | XP_016873964.1 | ||||
XM_017018476.1 | Intron | XP_016873965.1 |