Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601618 MIM: 604784 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SOX18 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
SOX18 - SRY-box 18 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018419.2 | Intron | NP_060889.1 |
TCEA2 - transcription elongation factor A2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003195.4 | Intron | NP_003186.1 | ||||
NM_198723.1 | Intron | NP_942016.1 | ||||
XM_005260229.3 | Intron | XP_005260286.1 | ||||
XM_011529022.1 | Intron | XP_011527324.1 | ||||
XM_011529023.1 | Intron | XP_011527325.1 | ||||
XM_011529027.2 | Intron | XP_011527329.1 | ||||
XM_017028036.1 | Intron | XP_016883525.1 | ||||
XM_017028037.1 | Intron | XP_016883526.1 |
Set Membership: |
HapMap |