Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601191 | ||||||||||||||||||||
Literature Links: |
ERH PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ERH - enhancer of rudimentary homolog (Drosophila) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC39A9 - solute carrier family 39 member 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001252148.1 | Intron | NP_001239077.1 | ||||
NM_001252150.1 | Intron | NP_001239079.1 | ||||
NM_001252151.1 | Intron | NP_001239080.1 | ||||
NM_001252152.1 | Intron | NP_001239081.1 | ||||
NM_018375.4 | Intron | NP_060845.2 | ||||
XM_005267834.1 | Intron | XP_005267891.1 | ||||
XM_017021426.1 | Intron | XP_016876915.1 |