Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGACTCCTTTCATTTGATGACCATC[C/T]TAAGATCATAAGAGAAGCTTTTTTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM181A PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
FAM181A - family with sequence similarity 181 member A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001207071.1 | Intron | NP_001194000.1 | ||||
NM_001207072.1 | Intron | NP_001194001.1 | ||||
NM_001207073.1 | Intron | NP_001194002.1 | ||||
NM_001207074.1 | Intron | NP_001194003.1 | ||||
NM_138344.4 | Intron | NP_612353.3 | ||||
XM_011537297.1 | Intron | XP_011535599.1 |
FAM181A-AS1 - FAM181A antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |