Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616790 MIM: 605271 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PPP4R4 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PPP4R4 - protein phosphatase 4 regulatory subunit 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SERPINA10 - serpin family A member 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001100607.2 | 3708 | Intron | NP_001094077.1 | |||
NM_016186.2 | 3708 | Intron | NP_057270.1 | |||
XM_005267733.4 | 3708 | UTR 3 | XP_005267790.1 | |||
XM_017021353.1 | 3708 | UTR 3 | XP_016876842.1 |
Set Membership: |
HapMap |