Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DENND2A PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
DENND2A - DENN domain containing 2A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318052.1 | Intron | NP_001304981.1 | ||||
NM_001318053.1 | Intron | NP_001304982.1 | ||||
NM_015689.4 | Intron | NP_056504.3 | ||||
XM_005249977.3 | Intron | XP_005250034.1 | ||||
XM_005249979.4 | Intron | XP_005250036.1 | ||||
XM_011516052.2 | Intron | XP_011514354.1 | ||||
XM_011516053.2 | Intron | XP_011514355.1 | ||||
XM_011516054.2 | Intron | XP_011514356.1 | ||||
XM_017011989.1 | Intron | XP_016867478.1 |