Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611061 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM20C PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FAM20C - family with sequence similarity 20 member C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020223.3 | 35 | Intron | NP_064608.2 | |||
XM_017012450.1 | 35 | Intron | XP_016867939.1 | |||
XM_017012451.1 | 35 | Intron | XP_016867940.1 | |||
XM_017012452.1 | 35 | Intron | XP_016867941.1 | |||
XM_017012453.1 | 35 | Intron | XP_016867942.1 | |||
XM_017012454.1 | 35 | Intron | XP_016867943.1 | |||
XM_017012455.1 | 35 | UTR 5 | XP_016867944.1 |