Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602351 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CMKLR1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CMKLR1 - chemerin chemokine-like receptor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142343.1 | 2325 | UTR 3 | NP_001135815.1 | |||
NM_001142344.1 | 2325 | UTR 3 | NP_001135816.1 | |||
NM_001142345.1 | 2325 | UTR 3 | NP_001135817.1 | |||
NM_004072.2 | 2325 | UTR 3 | NP_004063.1 | |||
XM_017018820.1 | 2325 | UTR 3 | XP_016874309.1 |
Set Membership: |
HapMap |