Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609445 MIM: 606202 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RXFP3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
RXFP3 - relaxin/insulin like family peptide receptor 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC45A2 - solute carrier family 45 member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001012509.3 | 2173 | UTR 3 | NP_001012527.1 | |||
NM_001297417.2 | 2173 | Intron | NP_001284346.2 | |||
NM_016180.4 | 2173 | Intron | NP_057264.3 |
Set Membership: |
HapMap |