Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615124 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ANKRD13B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ANKRD13B - ankyrin repeat domain 13B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152345.4 | 2292 | Intron | NP_689558.4 | |||
XM_005257920.4 | 2292 | Intron | XP_005257977.1 | |||
XM_011524319.2 | 2292 | Intron | XP_011522621.1 | |||
XM_011524320.2 | 2292 | Intron | XP_011522622.1 | |||
XM_011524321.2 | 2292 | Intron | XP_011522623.1 | |||
XM_017024174.1 | 2292 | Intron | XP_016879663.1 | |||
XM_017024175.1 | 2292 | Intron | XP_016879664.1 | |||
XM_017024176.1 | 2292 | Intron | XP_016879665.1 |
CORO6 - coronin 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032854.3 | 2292 | UTR 3 | NP_116243.2 | |||
XM_005258047.3 | 2292 | UTR 3 | XP_005258104.1 | |||
XM_005258048.4 | 2292 | UTR 3 | XP_005258105.2 | |||
XM_005258056.4 | 2292 | UTR 3 | XP_005258113.1 | |||
XM_011525385.2 | 2292 | UTR 3 | XP_011523687.1 | |||
XM_011525386.2 | 2292 | UTR 3 | XP_011523688.1 | |||
XM_011525387.2 | 2292 | UTR 3 | XP_011523689.1 | |||
XM_011525388.2 | 2292 | Intron | XP_011523690.1 | |||
XM_011525389.2 | 2292 | Intron | XP_011523691.1 | |||
XM_011525390.2 | 2292 | UTR 3 | XP_011523692.1 | |||
XM_011525391.2 | 2292 | UTR 3 | XP_011523693.1 | |||
XM_017025235.1 | 2292 | UTR 3 | XP_016880724.1 |