Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603016 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CDH19 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CDH19 - cadherin 19 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271028.1 | 4162 | UTR 3 | NP_001257957.1 | |||
NM_021153.3 | 4162 | UTR 3 | NP_066976.1 | |||
XM_011525931.2 | 4162 | UTR 3 | XP_011524233.1 | |||
XM_011525932.1 | 4162 | Intron | XP_011524234.1 | |||
XM_017025711.1 | 4162 | UTR 3 | XP_016881200.1 |
Set Membership: |
HapMap |