Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608467 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
STON2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
STON2 - stonin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256430.1 | 12420 | UTR 3 | NP_001243359.1 | |||
NM_033104.3 | 12420 | UTR 3 | NP_149095.2 | |||
XM_006720279.1 | 12420 | UTR 3 | XP_006720342.1 | |||
XM_011537240.1 | 12420 | UTR 3 | XP_011535542.1 | |||
XM_011537241.1 | 12420 | Intron | XP_011535543.1 | |||
XM_011537242.1 | 12420 | UTR 3 | XP_011535544.1 |
Set Membership: |
HapMap |