Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604143 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ESPL1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ESPL1 - extra spindle pole bodies like 1, separase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012291.4 | 165 | Missense Mutation | GAC,GCC | D,A 25 | NP_036423.4 | |
XM_006719705.3 | 165 | Missense Mutation | GAC,GCC | D,A 25 | XP_006719768.1 | |
XM_011539024.2 | 165 | Missense Mutation | GAC,GCC | D,A 25 | XP_011537326.1 | |
XM_011539025.2 | 165 | Missense Mutation | GAC,GCC | D,A 25 | XP_011537327.1 | |
XM_017020253.1 | 165 | UTR 5 | XP_016875742.1 |
Set Membership: |
HapMap |