Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608272 MIM: 606107 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NEU1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
NEU1 - neuraminidase 1 (lysosomal sialidase) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC44A4 - solute carrier family 44 member 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001178044.1 | 1546 | Missense Mutation | CGC,TGC | R,C 451 | NP_001171515.1 | |
NM_001178045.1 | 1546 | Missense Mutation | CGC,TGC | R,C 417 | NP_001171516.1 | |
NM_025257.2 | 1546 | Missense Mutation | CGC,TGC | R,C 493 | NP_079533.2 |
Set Membership: |
HapMap |