Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTGGGAGGTCAAGGCCAGCTATAT[A/T]TGAGCCCTTTTGAGAAGGACATAGG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609991 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FNDC1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
FNDC1 - fibronectin type III domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032532.2 | Intron | NP_115921.2 | ||||
XM_011536190.2 | Intron | XP_011534492.1 | ||||
XM_011536191.2 | Intron | XP_011534493.1 |
LOC101929122 - uncharacterized LOC101929122 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |