Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608188 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CRLS1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CRLS1 - cardiolipin synthase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001127458.1 | Intron | NP_001120930.1 | ||||
NM_001323561.1 | Intron | NP_001310490.1 | ||||
NM_001323562.1 | Intron | NP_001310491.1 | ||||
NM_001323563.1 | Intron | NP_001310492.1 | ||||
NM_001323564.1 | Intron | NP_001310493.1 | ||||
NM_019095.5 | Intron | NP_061968.1 | ||||
XM_005260738.4 | Intron | XP_005260795.1 | ||||
XM_011529263.1 | Intron | XP_011527565.1 |
Set Membership: |
HapMap |