Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616650 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KIAA0556 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
KIAA0556 - KIAA0556 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015202.3 | Intron | NP_056017.3 | ||||
XM_005255201.3 | Intron | XP_005255258.1 | ||||
XM_005255202.3 | Intron | XP_005255259.1 | ||||
XM_005255203.3 | Intron | XP_005255260.1 | ||||
XM_005255206.3 | Intron | XP_005255263.1 | ||||
XM_005255208.2 | Intron | XP_005255265.1 | ||||
XM_006721025.3 | Intron | XP_006721088.1 | ||||
XM_011545773.2 | Intron | XP_011544075.1 | ||||
XM_011545774.1 | Intron | XP_011544076.1 | ||||
XM_011545775.2 | Intron | XP_011544077.1 | ||||
XM_011545776.2 | Intron | XP_011544078.1 | ||||
XM_011545777.2 | Intron | XP_011544079.1 | ||||
XM_017023085.1 | Intron | XP_016878574.1 | ||||
XM_017023086.1 | Intron | XP_016878575.1 | ||||
XM_017023087.1 | Intron | XP_016878576.1 | ||||
XM_017023088.1 | Intron | XP_016878577.1 |