Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610490 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ACP7 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ACP7 - acid phosphatase 7, tartrate resistant (putative) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001004318.2 | 115 | UTR 5 | NP_001004318.2 | |||
XM_011526966.2 | 115 | UTR 5 | XP_011525268.1 | |||
XM_011526967.2 | 115 | UTR 5 | XP_011525269.1 | |||
XM_011526969.1 | 115 | UTR 5 | XP_011525271.1 | |||
XM_017026807.1 | 115 | UTR 5 | XP_016882296.1 | |||
XM_017026808.1 | 115 | UTR 5 | XP_016882297.1 |
Set Membership: |
HapMap |