Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610907 MIM: 601844 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
VPS25 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
VPS25 - vacuolar protein sorting 25 homolog | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
WNK4 - WNK lysine deficient protein kinase 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321299.1 | Intron | NP_001308228.1 | ||||
NM_032387.4 | Intron | NP_115763.2 | ||||
XM_011525135.2 | Intron | XP_011523437.1 | ||||
XM_017024962.1 | Intron | XP_016880451.1 | ||||
XM_017024963.1 | Intron | XP_016880452.1 | ||||
XM_017024964.1 | Intron | XP_016880453.1 | ||||
XM_017024965.1 | Intron | XP_016880454.1 | ||||
XM_017024966.1 | Intron | XP_016880455.1 |
Set Membership: |
HapMap |