Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605696 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KCNG2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KCNG2 - potassium voltage-gated channel modifier subfamily G member 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PQLC1 - PQ loop repeat containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001146343.1 | Intron | NP_001139815.1 | ||||
NM_001146345.1 | Intron | NP_001139817.1 | ||||
NM_025078.4 | Intron | NP_079354.2 | ||||
XM_005266770.2 | Intron | XP_005266827.1 | ||||
XM_011526188.1 | Intron | XP_011524490.1 | ||||
XM_017025997.1 | Intron | XP_016881486.1 | ||||
XM_017025998.1 | Intron | XP_016881487.1 | ||||
XM_017025999.1 | Intron | XP_016881488.1 | ||||
XM_017026000.1 | Intron | XP_016881489.1 | ||||
XM_017026001.1 | Intron | XP_016881490.1 | ||||
XM_017026002.1 | Intron | XP_016881491.1 | ||||
XM_017026003.1 | Intron | XP_016881492.1 | ||||
XM_017026004.1 | Intron | XP_016881493.1 | ||||
XM_017026005.1 | Intron | XP_016881494.1 |