Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600902 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SEPHS1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SEPHS1 - selenophosphate synthetase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001195602.1 | 1137 | Intron | NP_001182531.1 | |||
NM_001195604.1 | 1137 | Intron | NP_001182533.1 | |||
NM_012247.4 | 1137 | Intron | NP_036379.2 | |||
XM_017015942.1 | 1137 | Silent Mutation | ACC,ACG | T,T 257 | XP_016871431.1 | |
XM_017015943.1 | 1137 | Silent Mutation | ACC,ACG | T,T 257 | XP_016871432.1 | |
XM_017015944.1 | 1137 | Silent Mutation | ACC,ACG | T,T 255 | XP_016871433.1 | |
XM_017015945.1 | 1137 | Intron | XP_016871434.1 |