Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604602 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MRRF PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
MRRF - mitochondrial ribosome recycling factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001173512.1 | 238 | UTR 5 | NP_001166983.1 | |||
NM_138777.3 | 238 | UTR 5 | NP_620132.1 | |||
NM_199177.2 | 238 | Intron | NP_954646.1 | |||
XM_005252302.3 | 238 | Intron | XP_005252359.1 | |||
XM_011519183.2 | 238 | Intron | XP_011517485.1 | |||
XM_011519184.2 | 238 | Intron | XP_011517486.1 | |||
XM_011519185.2 | 238 | Intron | XP_011517487.1 | |||
XM_011519186.1 | 238 | Intron | XP_011517488.1 | |||
XM_011519187.2 | 238 | UTR 5 | XP_011517489.1 | |||
XM_017015286.1 | 238 | Intron | XP_016870775.1 | |||
XM_017015287.1 | 238 | Intron | XP_016870776.1 |
RBM18 - RNA binding motif protein 18 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_033117.3 | 238 | UTR 5 | NP_149108.1 | |||
XM_006717319.1 | 238 | Intron | XP_006717382.1 |
Set Membership: |
HapMap |