Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTGTGTTAGGTACAGGGGTCACTT[A/C]AAATGTAGCTAATCCAAACAAATGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606127 | ||||||||||||||||||||
Literature Links: |
LOC101928418 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LOC101928418 - uncharacterized LOC101928418 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MYOCD - myocardin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001146312.2 | Intron | NP_001139784.1 | ||||
NM_153604.3 | Intron | NP_705832.1 | ||||
XM_005256863.1 | Intron | XP_005256920.1 | ||||
XM_005256864.1 | Intron | XP_005256921.1 | ||||
XM_017025342.1 | Intron | XP_016880831.1 |