Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SNHG25 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SNHG25 - small nucleolar RNA host gene 25 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNORA50C - small nucleolar RNA, H/ACA box 50C | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNORD104 - small nucleolar RNA, C/D box 104 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TEX2 - testis expressed 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001288732.1 | Intron | NP_001275661.1 | ||||
NM_001288733.1 | Intron | NP_001275662.1 | ||||
NM_018469.4 | Intron | NP_060939.3 | ||||
XM_011524998.1 | Intron | XP_011523300.1 | ||||
XM_011524999.1 | Intron | XP_011523301.1 | ||||
XM_011525000.1 | Intron | XP_011523302.1 | ||||
XM_017024846.1 | Intron | XP_016880335.1 | ||||
XM_017024847.1 | Intron | XP_016880336.1 |