Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604511 MIM: 614906 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CBX1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CBX1 - chromobox 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNX11 - sorting nexin 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_013323.2 | Intron | NP_037455.2 | ||||
NM_152244.1 | Intron | NP_689450.1 | ||||
XM_005257260.3 | Intron | XP_005257317.1 | ||||
XM_005257261.3 | Intron | XP_005257318.1 | ||||
XM_005257262.3 | Intron | XP_005257319.1 | ||||
XM_011524697.2 | Intron | XP_011522999.1 |