Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603248 MIM: 603610 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
BMPR1B PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
BMPR1B - bone morphogenetic protein receptor type 1B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
UNC5C - unc-5 netrin receptor C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003728.3 | 8485 | UTR 3 | NP_003719.3 | |||
XM_005263321.3 | 8485 | UTR 3 | XP_005263378.1 | |||
XM_017008789.1 | 8485 | UTR 3 | XP_016864278.1 | |||
XM_017008790.1 | 8485 | UTR 3 | XP_016864279.1 |
Set Membership: |
HapMap |