Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCGGGCCTGCCACAATTAAATGAC[C/G]GCAAGAAAGAAGTCTATGCTAAGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LYPD5 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LYPD5 - LY6/PLAUR domain containing 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001031749.2 | 196 | Intron | NP_001026919.2 | |||
NM_001288763.1 | 196 | Intron | NP_001275692.1 | |||
NM_182573.2 | 196 | Intron | NP_872379.2 | |||
XM_011526794.2 | 196 | Intron | XP_011525096.1 |
ZNF283 - zinc finger protein 283 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001297752.1 | 196 | UTR 5 | NP_001284681.1 | |||
NM_181845.1 | 196 | Intron | NP_862828.1 | |||
XM_005258784.3 | 196 | UTR 5 | XP_005258841.1 | |||
XM_005258785.3 | 196 | Intron | XP_005258842.1 | |||
XM_005258786.3 | 196 | UTR 5 | XP_005258843.1 | |||
XM_017026628.1 | 196 | UTR 5 | XP_016882117.1 | |||
XM_017026629.1 | 196 | UTR 5 | XP_016882118.1 | |||
XM_017026630.1 | 196 | Intron | XP_016882119.1 | |||
XM_017026631.1 | 196 | Intron | XP_016882120.1 | |||
XM_017026632.1 | 196 | UTR 5 | XP_016882121.1 | |||
XM_017026633.1 | 196 | UTR 5 | XP_016882122.1 | |||
XM_017026634.1 | 196 | Intron | XP_016882123.1 | |||
XM_017026635.1 | 196 | UTR 5 | XP_016882124.1 | |||
XM_017026636.1 | 196 | Intron | XP_016882125.1 | |||
XM_017026637.1 | 196 | UTR 5 | XP_016882126.1 | |||
XM_017026638.1 | 196 | Intron | XP_016882127.1 | |||
XM_017026639.1 | 196 | UTR 5 | XP_016882128.1 |
Set Membership: |
HapMap |