Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACTAAAAATTGTAAGGATTTATAT[C/T]CAAGTGATAGATCATTCTAAGAACT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607193 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RGS20 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
RGS20 - regulator of G-protein signaling 20 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286673.1 | Intron | NP_001273602.1 | ||||
NM_001286674.1 | Intron | NP_001273603.1 | ||||
NM_001286675.1 | Intron | NP_001273604.1 | ||||
NM_003702.4 | Intron | NP_003693.2 | ||||
NM_170587.3 | Intron | NP_733466.1 | ||||
XM_011517622.2 | Intron | XP_011515924.1 |
Set Membership: |
HapMap |