Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATGATGAGATCTCTCTAAATAGAC[A/G]TAATTATCAGGTGAAGATTTAAGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CWC27 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CWC27 - CWC27 spliceosome associated protein homolog | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001297644.1 | Intron | NP_001284573.1 | ||||
NM_001297645.1 | Intron | NP_001284574.1 | ||||
NM_001318000.1 | Intron | NP_001304929.1 | ||||
NM_005869.3 | Intron | NP_005860.2 | ||||
XM_011543095.2 | Intron | XP_011541397.2 | ||||
XM_011543096.1 | Intron | XP_011541398.1 | ||||
XM_017008943.1 | Intron | XP_016864432.1 |
SREK1IP1 - SREK1 interacting protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |