Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTGGGCAACATAGCCAGACCTCCT[C/G]TCTACAAAAAGAAAAAAGAATGACA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
UBE2CP5 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
UBE2CP5 - ubiquitin conjugating enzyme E2 C pseudogene 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF587 - zinc finger protein 587 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001204817.1 | Intron | NP_001191746.1 | ||||
NM_032828.3 | Intron | NP_116217.1 |
ZNF814 - zinc finger protein 814 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |