Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTGTGTGTGTTTTCATATTTTATT[A/C]TTTCCCCCCCTCATCTTAGGGTCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC105377310 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LOC105377310 - syncytin-1-like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RPS6P6 - ribosomal protein S6 pseudogene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM150C - transmembrane protein 150C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080506.1 | Intron | NP_001073975.1 |
Set Membership: |
HapMap |