Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606238 MIM: 611310 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
IKZF5 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
IKZF5 - IKAROS family zinc finger 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271840.1 | 4327 | UTR 3 | NP_001258769.1 | |||
XM_006717947.3 | 4327 | UTR 3 | XP_006718010.1 | |||
XM_017016550.1 | 4327 | UTR 3 | XP_016872039.1 | |||
XM_017016551.1 | 4327 | UTR 3 | XP_016872040.1 | |||
XM_017016552.1 | 4327 | UTR 3 | XP_016872041.1 | |||
XM_017016553.1 | 4327 | UTR 3 | XP_016872042.1 | |||
XM_017016554.1 | 4327 | UTR 3 | XP_016872043.1 | |||
XM_017016555.1 | 4327 | UTR 3 | XP_016872044.1 |
PSTK - phosphoseryl-tRNA kinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_153336.2 | 4327 | Intron | NP_699167.2 | |||
XM_017015640.1 | 4327 | Intron | XP_016871129.1 | |||
XM_017015641.1 | 4327 | Intron | XP_016871130.1 |
Set Membership: |
HapMap |