Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTTCAGTCCTACTGGTGGCCCTCA[A/G]ATCGGTTACTCATTCAACCATCCAG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608876 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ANKRD42 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ANKRD42 - ankyrin repeat domain 42 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001300972.1 | Intron | NP_001287901.1 | ||||
NM_001300973.1 | Intron | NP_001287902.1 | ||||
NM_001300975.1 | Intron | NP_001287904.1 | ||||
NM_001300976.1 | Intron | NP_001287905.1 | ||||
NM_001300977.1 | Intron | NP_001287906.1 | ||||
NM_182603.3 | Intron | NP_872409.2 |
LOC100506282 - uncharacterized LOC100506282 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PCF11 - PCF11 cleavage and polyadenylation factor subunit | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PCF11-AS1 - PCF11 anstisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |