Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
33 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608042 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SYTL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SYTL1 - synaptotagmin like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001193308.1 | 204 | Intron | NP_001180237.1 | |||
NM_032872.2 | 204 | Intron | NP_116261.1 | |||
XM_005246022.3 | 204 | UTR 5 | XP_005246079.1 | |||
XM_006710990.1 | 204 | UTR 5 | XP_006711053.1 | |||
XM_006710991.1 | 204 | Intron | XP_006711054.1 |
TMEM222 - transmembrane protein 222 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |