Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAGGTTGATTTCTAGTTCTTCAGAG[A/G]TATTTTACAAGATAATTATATGTAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 102772 MIM: 616641 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
AMPD3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
AMPD3 - adenosine monophosphate deaminase 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR4485 - microRNA 4485 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MTRNR2L8 - MT-RNR2-like 8 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RNF141 - ring finger protein 141 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016422.3 | Intron | NP_057506.2 |