Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTGATACAAAATTGCAGAGAGTCC[C/T]TGAGAAGAGAGATAGGCAGCAACTT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300566 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LHFPL1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
LHFPL1 - lipoma HMGIC fusion partner-like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_178175.3 | Intron | NP_835469.1 | ||||
XM_011530943.2 | Intron | XP_011529245.1 | ||||
XM_011530944.2 | Intron | XP_011529246.1 | ||||
XM_011530946.2 | Intron | XP_011529248.1 | ||||
XM_017029485.1 | Intron | XP_016884974.1 |