Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614955 MIM: 614957 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLFN12 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
SLFN12 - schlafen family member 12 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLFN13 - schlafen family member 13 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_144682.5 | 8302 | UTR 3 | NP_653283.3 | |||
XM_005257922.2 | 8302 | Intron | XP_005257979.1 | |||
XM_011524383.2 | 8302 | Intron | XP_011522685.1 | |||
XM_017024232.1 | 8302 | Intron | XP_016879721.1 | |||
XM_017024233.1 | 8302 | Intron | XP_016879722.1 |