Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610652 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PDE10A PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
PDE10A - phosphodiesterase 10A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130690.2 | 5157 | UTR 3 | NP_001124162.1 | |||
NM_006661.3 | 5157 | UTR 3 | NP_006652.1 | |||
XM_006715321.3 | 5157 | UTR 3 | XP_006715384.1 | |||
XM_011535387.2 | 5157 | UTR 3 | XP_011533689.2 | |||
XM_011535388.2 | 5157 | UTR 3 | XP_011533690.1 | |||
XM_011535393.2 | 5157 | UTR 3 | XP_011533695.1 | |||
XM_017010194.1 | 5157 | UTR 3 | XP_016865683.1 | |||
XM_017010195.1 | 5157 | UTR 3 | XP_016865684.1 | |||
XM_017010196.1 | 5157 | UTR 3 | XP_016865685.1 | |||
XM_017010197.1 | 5157 | Intron | XP_016865686.1 |
Set Membership: |
HapMap |