Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGCACCATAATACAAATTTTTTTA[C/T]ATACAGTATCCATCTATAATGCTCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300479 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM9C PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
FAM9C - family with sequence similarity 9 member C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_174901.5 | Intron | NP_777561.1 | ||||
XM_005274460.2 | Intron | XP_005274517.1 | ||||
XM_017029321.1 | Intron | XP_016884810.1 |
LOC105373133 - uncharacterized LOC105373133 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |