Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CEP95 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CEP95 - centrosomal protein 95 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001316990.1 | 1836 | Missense Mutation | ATA,ATG | I,M 1 | NP_001303919.1 | |
NM_138363.2 | 1836 | Missense Mutation | ATA,ATG | I,M 165 | NP_612372.1 | |
XM_005257779.3 | 1836 | Missense Mutation | ATA,ATG | I,M 165 | XP_005257836.1 | |
XM_005257780.2 | 1836 | Missense Mutation | ATA,ATG | I,M 1 | XP_005257837.1 | |
XM_006722166.3 | 1836 | UTR 5 | XP_006722229.1 | |||
XM_011525458.2 | 1836 | Missense Mutation | ATA,ATG | I,M 165 | XP_011523760.1 | |
XM_017025287.1 | 1836 | Missense Mutation | ATA,ATG | I,M 1 | XP_016880776.1 | |
XM_017025288.1 | 1836 | UTR 5 | XP_016880777.1 |